Sulphuric acid 0.5 mol/l (1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
7102.1000EA 36.8 EUR
7102.1000 7102.9010 7102.9020
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Sulphuric acid
Formula: H₂SO₄
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II