Sodium methoxide 30% in methanol, Sigma-Aldrich®
Supplier: Merck
Danger
519952-1L 519952-2.5L 519952-25ML
SIAL519952-1LEA 24.1 EUR
SIAL519952-1L SIAL519952-2.5L SIAL519952-25ML
Sodium methoxide 30% in methanol, Sigma-Aldrich®
Sodium methoxide
Formula: CH₃NaO
Density: 0.97 g/cm³ (20 °C)
MDL Number: MFCD00012179
CAS Number: 124-41-4
EINECS: 204-699-5
UN: 1289
ADR: 3,III