Methanesulphonic acid ≥70% in aqueous solution
Supplier: Roth Carl
Danger
4236.1 4236.2
ROTH4236.1EA 20.1 EUR
ROTH4236.1 ROTH4236.2
Methanesulphonic acid ≥70% in aqueous solution
Methanesulphonic acid
Formula: CH₃SO₃H
Storage Temperature: Ambient
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 200-898-6
UN: 2586
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebd3f86cbdbb894828247119a403c686b0ecddf&vid=01bb7db9690bd0b27a8ab4f277f8a301da72087f&prid=web&clid=SAPDAM