Potassium hydroxide standardised solution 0.1 N in aqueous solution
Supplier: Thermo Fisher Scientific
Warning
35599.K2 35599.M4
35599.K2EA 38.5 EUR
35599.K2 35599.M4
Potassium hydroxide standardised solution 0.1 N in aqueous solution
Potassium hydroxide
CAS No.: 1310-58-3 Molecular Formula: HKO Formula Weight: 56.11 Physical Form: Liquid Appearance: Colourless
Formula: KOH
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II