Hydrochloric acid 25%
Supplier: Roth Carl
Danger
Synonyms: Hydrochloric acid solution, Muriatic acid
6331.1 6331.2 6331.3 6331.4
ROTH6331.1EA 0 EUR
ROTH6331.1 ROTH6331.2 ROTH6331.3 ROTH6331.4
Hydrochloric acid 25%
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II