Hydrochloric acid 32%
Supplier: Roth Carl
Danger
Synonyms: Hydrochloric acid solution, Muriatic acid
X896.1 X896.2
ROTHX896.1EA 23.2 EUR
ROTHX896.1 ROTHX896.2
Hydrochloric acid 32%
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II