Hydrochloric acid 5% v/v aqueous solution
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Hydrochloric acid solution, Muriatic acid
43335.K2 43335.K7 43335.AP
43335.K2EA 36.9 EUR
43335.K2 43335.K7 43335.AP
Hydrochloric acid 5% v/v aqueous solution
Hydrochloric acid
Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colourless to pale yellow UN No.: 1789 MDL No.: MFCD00011324
Formula: HCl
MW: 36.46 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.02…1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III