Phosphorus(III) bromide 1 mol/l in dichloromethane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Phosphorus tribromide
M-Clarity™ quality level = MQ100
276820-100ML 276820-800ML
SIAL276820-100MLEA 67.5 EUR
SIAL276820-100ML SIAL276820-800ML
Phosphorus(III) bromide 1 mol/l in dichloromethane, Sigma-Aldrich®
Phosphorus (III) bromide
Formula: PBr₃
MW: 270.69 g/mol
Boiling Pt: 175
Melting Pt: -40
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
Merck Index: 12,07514