Sodium hydroxide 30% (w/w) 40% (w/v) in aqueous solution for Kjeldahl determinations
Danger
Synonyms: Caustic soda
S30WWLN S30WW1 S30WW5
REGES30WWLNEA 130 EUR
REGES30WWLN REGES30WW1 REGES30WW5
Sodium hydroxide 30% (w/w) 40% (w/v) in aqueous solution for Kjeldahl determinations
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II