Tetrabutylammonium fluoride ≥75% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
B21261.14 B21261.22 B21261.36
B21261.14EA 59.1 EUR
B21261.14 B21261.22 B21261.36
Tetrabutylammonium fluoride ≥75% (w/w) in aqueous solution
Tetrabutylammonium fluoride
CAS No.: 429-41-4 Molecular Formula: [CH3(CH2)3]4NF Formula Weight: 261.47 UN No.: 2922 MDL No.: MFCD00011747
Formula: C₁₆H₃₆FN
MW: 261.47 g/mol
Density: 0.903
Storage Temperature: Ambient
MDL Number: MFCD00011747
CAS Number: 429-41-4
EINECS: 207-057-2
UN: 2922
ADR: 8 II