Tetraethylammonium hydroxide 35% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Tetrylammonium hydroxide
36560.22
36560.36
36560.A3
36560.22EA
79.8
EUR
36560.22
36560.36
36560.A3
Tetraethylammonium hydroxide 35% (w/w) in aqueous solution
Tetraethylammonium hydroxide
CAS No.: 77-98-5
Molecular Formula: C8H21NO
Formula Weight: 147.26
Physical Form: Liquid
Density: 1.023
Refractive Index: 1.4040
Merck Reference: 14,9201
UN No.: 3267
MDL No.: MFCD00009024
Formula:
C₈H₂₁NO MW: 147.26 g/mol Density: 1.023 Storage Temperature: Ambient |
MDL Number:
MFCD00009024 CAS Number: 77-98-5 EINECS: 201-073-3 UN: 3267 ADR: 8 II Merck Index: 14,9201 |