Tetramethylammonium hydroxide 5% in aqueous solution
Supplier: VWR Chemicals
28561.187EA
0
EUR
28561.187
Tetramethylammonium hydroxide 5% in aqueous solution
Tetramethylammonium hydroxide
Formula:
C₄H₁₃NO MW: 91,15 g/mol Boiling Pt: 110 °C (1013 hPa) Density: 1,02 g/cm³ (20 °C) |
MDL Number:
MFCD00008280 UN: 1835 ADR: 8,II |
Specification Test Results
Assay | Min. 5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...