L(+)-Cysteine hydrochloride monohydrate, TECHNICAL
Supplier: VWR Chemicals
23255.186EA 60.1 EUR
23255.186
L(+)-Cysteine hydrochloride monohydrate, TECHNICAL
L(+)-Cysteine hydrochloride monohydrate
Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175,64 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD00065606
CAS Number: 7048-04-6
Merck Index: 13,02810
https://si.vwr.com/stibo/web/std.lang.all/58/37/2345837.jpg
Order Now
Specification Test Results
Identification Passes test