Spermidine trihydrochloride 99+%
Supplier: Thermo Scientific
Warning
Synonyms: 4-Azoniaoctamethylenediammonium trichloride, 4-Azaoctamethylenediamine trihydrochloride, N1-(3-Aminopropyl)butane-1,4-diamine trihydrochloride, N-(3-Aminopropyl)-1,4-diaminobutane trihydrochloride
215100050 215100010 215100250
ACRO215100050EA 115 EUR
ACRO215100050 ACRO215100010 ACRO215100250
Spermidine trihydrochloride 99+%
Spermidine trihydrochloride
Formula: C₇H₂₂Cl₃N₃
MW: 254.63 g/mol
Melting Pt: 248…252 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012918
CAS Number: 334-50-9
EINECS: 206-379-0
Merck Index: 12,08893
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %