N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: N1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochloride, TMPD 2 HCl, N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochloride, N,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride
M-Clarity™ quality level = MQ200
87890-25G 87890-5G
SIAL87890-25GEA 122 EUR
SIAL87890-25G SIAL87890-5G
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, Sigma-Aldrich®
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Formula: C₁₀H₁₈Cl₂N₂
MW: 237.17 g/mol
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00012482
CAS Number: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e955f6b2a1434ddba239f533c6611583219f8904&vid=13fb110b013db70c026f8eefeee7f08fa2568c0c&prid=web&clid=SAPDAM