Guanidinium hydrochloride, Sigma-Aldrich®
Supplier: Merck
Warning
50933-1KG 50933-250G 50933-50G
SIAL50933-1KGEA 2940 EUR
SIAL50933-1KG SIAL50933-250G SIAL50933-50G
Guanidinium hydrochloride, Sigma-Aldrich®
Guanidinium hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
Melting Pt: 185 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM