Guanidinium hydrochloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
50950-1KG 50950-2.5KG 50950-250G
SIAL50950-1KGEA 146 EUR
SIAL50950-1KG SIAL50950-2.5KG SIAL50950-250G
Guanidinium hydrochloride, Sigma-Aldrich®
Guanidinium hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM