Guanidinium thiocyanate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
50980-100G 50980-250G 50980-25G
SIAL50980-100GEA 464 EUR
SIAL50980-100G SIAL50980-250G SIAL50980-25G
Guanidinium thiocyanate, Sigma-Aldrich®
Guanidinium thiocyanate
Formula: CH₅N₃·CHNS
Melting Pt: 118 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1