Hydroxylammonium chloride 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

A15398.0B A15398.0I A15398.22 A15398.36
A15398.0BEA 148 EUR
A15398.0B A15398.0I A15398.22 A15398.36
Hydroxylammonium chloride 99%
Hydroxylammonium chloride

Purity: 99% CAS No.: 5470-11-1 Molecular Formula: H3NO.HCl Formula Weight: 69.49 Odour: Acidic pH: 3.6 Melting Point: 152° dec. Beilstein Reference: 3539763 Merck Reference: 14,4828 UN No.: 2923 MDL No.: MFCD00051089

Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306
Melting Pt: 152 °C (dec.)
Density: 1.67
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8 III
Merck Index: 13,04853

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR