Hydroxylammonium chloride, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

M-Clarity™ quality level = MQ200

379921-25G 379921-5G
SIAL379921-25GEA 110 EUR
SIAL379921-25G SIAL379921-5G
Hydroxylammonium chloride, Sigma-Aldrich®
Hydroxylammonium chloride
Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306
Melting Pt: 151
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8 III
Merck Index: 13,04853

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR