Hydrazinium sulphate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Hydrazine sulphate salt, Hydrazine sulphate

M-Clarity™ quality level = MQ200

216046-100G 216046-500G 216046-5G
SIAL216046-100GEA 49.7 EUR
SIAL216046-100G SIAL216046-500G SIAL216046-5G
Hydrazinium sulphate, Sigma-Aldrich®
Hydrazinium sulphate
Formula: NH₂NH₂·H₂SO₄
MW: 130.12 g/mol
Melting Pt: 254 °C
Density: 1.37 g/cm³ (20 °C)
MDL Number: MFCD00044873
CAS Number: 10034-93-2
EINECS: 233-110-4
UN: 3288
ADR: 6.1,III
Merck Index: 14,04772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e277940036b3cf71839ff7458f0c7d08bff6f6d5&vid=7a677e6f1725cfa1b9bdef05cc04304e6739079e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR