Hydrazinium sulphate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Hydrazine sulphate salt, Hydrazine sulphate
53900-100G
53900-500G
SIAL53900-100GEA
44.3
EUR
SIAL53900-100G
SIAL53900-500G
Hydrazinium sulphate, Sigma-Aldrich®
Hydrazinium sulphate
Formula:
NH₂NH₂·H₂SO₄ MW: 130.12 g/mol Melting Pt: 254 °C Density: 1.37 g/cm³ (20 °C) |
MDL Number:
MFCD00044873 CAS Number: 10034-93-2 EINECS: 233-110-4 UN: 3288 ADR: 6.1,III Merck Index: 14,04772 |
|
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...