Hydrazinium monochloride 98+%
Supplier: Thermo Scientific
Danger
Synonyms: Hydrazinium monochloride, Hydrazinium chloride, Hydrazine monohydrochloride, Hydrazine hydrochloride
201990010 201992500 201990051
ACRO201990010EA 235 EUR
ACRO201990010 ACRO201992500 ACRO201990051
Hydrazinium monochloride 98+%
Hydrazinium monochloride
Formula: H₅ClN₂
MW: 68.51 g/mol
Melting Pt: 89…93 °C
Density: 1,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044368
CAS Number: 2644-70-4
EINECS: 220-154-4
UN: 3288
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥98.0 %