Hydrazine dihydrochloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Hydrazine Dihydrochloride, Hydrazinium dichloride

M-Clarity™ quality level = MQ100

450251-25G 450251-5G
SIAL450251-25GEA 61.1 EUR
SIAL450251-25G SIAL450251-5G
Hydrazine dihydrochloride, Sigma-Aldrich®
Hydrazine dihydrochloride
Formula: H₆Cl₂N₂
MW: 104.97 g/mol
Melting Pt: 189 °C
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00064543
CAS Number: 5341-61-7
EINECS: 226-283-2
UN: 2928
ADR: 6.1,II
Merck Index: 13,04789

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR