Phenylhydrazinium chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL114715-500GEA 245 EUR
SIAL114715-500G SAFA114715-5G
Phenylhydrazinium chloride, Sigma-Aldrich®
Phenylhydrazinium chloride
Formula: C₆H₉ClN₂
MW: 144,6 g/mol
Melting Pt: 243…246 °C
MDL Number: MFCD00012924
UN: 2811
ADR: 6.1,III
Merck Index: 12,07447
https://si.vwr.com/stibo/web/std.lang.all/74/26/2297426.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR