N-(1-Naphthyl)ethylenediamine dihydrochloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine
N9125-100G N9125-10G N9125-25G
SIALN9125-100GEA 320 EUR
SIALN9125-100G SIALN9125-10G SIALN9125-25G
N-(1-Naphthyl)ethylenediamine dihydrochloride, Sigma-Aldrich®
N-2-Aminoethyl-1-naphthylamine dihydrochloride
Formula: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS Number: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM