Sodium acetate trihydrate
Supplier: Roth Carl
Synonyms: Acetic acid sodium salt trihydrate
3856.1 3856.2 3856.5
ROTH3856.1EA 26.7 EUR
ROTH3856.1 ROTH3856.2 ROTH3856.5
Sodium acetate trihydrate
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM