Sodium acetate trihydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
S1304-1KG
S1304-500G
S1304-5KG
SIALS1304-1KGEA
173
EUR
SIALS1304-1KG
SIALS1304-500G
SIALS1304-5KG
Sodium acetate trihydrate, Sigma-Aldrich®
Sodium acetate trihydrate
Formula:
H₃CCOONa·3H₂O Boiling Pt: 120…123 °C (1013 hPa) Melting Pt: 58 °C Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 EINECS: 204-823-8 Merck Index: 13,08642 |