Sodium acetate trihydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetic acid sodium salt trihydrate
MQ200
S7670-1KG S7670-250G S7670-500G S7670-5KG
SIALS7670-1KGEA 0 EUR
SIALS7670-1KG SIALS7670-250G SIALS7670-500G SIALS7670-5KG
Sodium acetate trihydrate, Sigma-Aldrich®
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM