Magnesium acetate tetrahydrate
Supplier: Roth Carl
Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate
0275.1 0275.2 0275.3
ROTH0275.1EA 37 EUR
ROTH0275.1 ROTH0275.2 ROTH0275.3
Magnesium acetate tetrahydrate
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80
Density: 1.454 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676