Magnesium acetate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate
M-Clarity™ quality level = MQ200
M2545-1KG M2545-250G
SIALM2545-1KGEA 350 EUR
SIALM2545-1KG SIALM2545-250G
Magnesium acetate tetrahydrate, Sigma-Aldrich®
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
Melting Pt: 80 °C
Density: 1.454 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676