Magnesium acetate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate
MQ200
M5661-250G M5661-50G
SIALM5661-250GEA 93.4 EUR
SIALM5661-250G SIALM5661-50G
Magnesium acetate tetrahydrate, Sigma-Aldrich®
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80
Density: 1.454 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676