Copper(II) acetate monohydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate
M-Clarity™ quality level = MQ200
229601-10G 229601-50G
SIAL229601-10GEA 205 EUR
SIAL229601-10G SIAL229601-50G
Copper(II) acetate monohydrate, Sigma-Aldrich®
Copper (II) acetate monohydrate
Formula: (H₃CCOO)₂Cu·H₂O
Melting Pt: 115 °C
Density: 1.88 g/cm³ (20 °C)
MDL Number: MFCD00149570
CAS Number: 6046-93-1
UN: 3077
ADR: 9,III
Merck Index: 13,02651