Manganese(II) acetate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Manganese acetate tetrahydrate, Manganese diacetate tetrahydrate, Acetic acid manganese (II) salt tetrahydrate
MQ200
221007-500G 221007-25G 221007-2KG
SIAL221007-500GEA 39.5 EUR
SIAL221007-500G SIAL221007-25G SIAL221007-2KG
Manganese(II) acetate tetrahydrate, Sigma-Aldrich®
Manganese (II) acetate tetrahydrate
Formula: (H₃CCOO)₂Mn·4H₂O
MW: 245.09 g/mol
Melting Pt: 300
Density: 1.58 g/cm³ (20 °C)
MDL Number: MFCD00062552
CAS Number: 6156-78-1
EINECS: 211-334-3
Merck Index: 13,05746