Oxalic acid dihydrate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Warning
Synonyms: Ethanedioic acid dihydrate
20555.296EA 50.4 EUR
20555.296 20555.365
Oxalic acid dihydrate ≥98%, TECHNICAL
Oxalic acid dihydrate
Formula: HO₂CCO₂H·2H₂O
Boiling Pt: 149…160 °C (1013 hPa)
Melting Pt: 101 °C
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8,III
REACH: 01-2119534576-33
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay Min. 98 %