tri-Sodium citrate dihydrate, Sigma-Aldrich®
Supplier: Merck
71402-1KG
71402-100G
71402-250G
SIAL71402-1KGEA
143
EUR
SIAL71402-1KG
SIAL71402-100G
SIAL71402-250G
tri-Sodium citrate dihydrate, Sigma-Aldrich®
tri-Sodium citrate dihydrate
Formula:
Na₃C₆H₅O₇·2H₂O Melting Pt: 150 °C Density: 1.76 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150031 CAS Number: 6132-04-3 EINECS: 200-675-3 Merck Index: 13,08675 |