tri-Sodium citrate dihydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
C7254-1KG
C7254-10KG
C7254-5KG
SIALC7254-1KGEA
41.8
EUR
SIALC7254-1KG
SIALC7254-10KG
SIALC7254-5KG
tri-Sodium citrate dihydrate, Sigma-Aldrich®
tri-Sodium citrate dihydrate
Formula:
Na₃C₆H₅O₇·2H₂O Melting Pt: 150 °C Density: 1.76 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150031 CAS Number: 6132-04-3 EINECS: 200-675-3 Merck Index: 13,08675 |