tri-Sodium citrate dihydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate
MQ200
C8532-100G C8532-10KG C8532-1KG C8532-500G C8532-5KG
SIALC8532-100GEA 44.1 EUR
SIALC8532-100G SIALC8532-10KG SIALC8532-1KG SIALC8532-500G SIALC8532-5KG
tri-Sodium citrate dihydrate, Sigma-Aldrich®
tri-Sodium citrate dihydrate
Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM