Stop solution 2
Supplier: Enzo Life Sciences
ENZOADI800377EA 48.4 EUR
ENZOADI800377 ALEXADI-80-0377 ENZOADI801804 ALEXADI-80-1804
Stop solution 2
Assays ELISAs
Acid stop solution (1 N HCl) intended for use as an assay stop reagent for colorimetric HRP/TMB reactions.
Each bottle is sufficient for use with 5×96 wells microtiter plates.
Informace pro objednávání: Additional assay development reagents are available as companion products to IMMUNOSET® products.