2,4,6-Trihydroxybenzoic acid monohydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2,4,6-Trihydroxybenzoic acid hydrate
M-Clarity™ quality level = MQ100
367346-100G
367346-25G
SIAL367346-100GEA
119
EUR
SIAL367346-100G
SIAL367346-25G
2,4,6-Trihydroxybenzoic acid monohydrate, Sigma-Aldrich®
2,4,6-Trihydroxybenzoic acid monohydrate
Formula:
C₇H₆O₅·1H₂O MW: 188.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149095 CAS Number: 71989-93-0 EINECS: 201-467-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...