p-Toluenesulphonic acid monohydrate
Supplier: Molekula
MOLE22682449-1KGEA
126
EUR
MOLE22682449-1KG
p-Toluenesulphonic acid monohydrate
p-Toluenesulphonic acid monohydrate
Formula:
C₇H₈O₃S·1H₂O MW: 190,22 g/mol Melting Pt: 102…104 °C Density: 1,24 g/cm³ (20 °C) Flash Pt: 180 °C |
MDL Number:
MFCD00142137 UN: 2585 ADR: 8,III Merck Index: 12,09671 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...