p-Toluenesulphonic acid monohydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate
M-Clarity™ quality level = MQ200
402885-100G 402885-500G 402885-5G
SIAL402885-100GEA 37.2 EUR
SIAL402885-100G SIAL402885-500G SIAL402885-5G
p-Toluenesulphonic acid monohydrate, Sigma-Aldrich®
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
MW: 190.22 g/mol
Melting Pt: 102…104 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8,III
Merck Index: 12,09671