tetra-Chloroauric(III) acid hydrate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate

50790-250MG 50790-1G 50790-5G
SIAL50790-250MGEA 89 EUR
SIAL50790-250MG SIAL50790-1G SIAL50790-5G
tetra-Chloroauric(III) acid hydrate, Sigma-Aldrich®
tetra-Chloroauric (III) acid hydrate
Formula: HAuCl₄·H₂O
MW: 357.8 g/mol
Density: 1.169 g/cm³ (20 °C)
MDL Number: MFCD00149903
CAS Number: 27988-77-8
EINECS: 240-948-4
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR