Potassium hexacyanoferrate(II) trihydrate

Supplier: Roth Carl

Synonyms: Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate

P745.1 P745.2
ROTHP745.1EA 113 EUR
ROTHP745.1 ROTHP745.2
Potassium hexacyanoferrate(II) trihydrate
Potassium hexacyanoferrate (II) trihydrate
Formula: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Melting Pt: 70
Density: 1.889 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR