J.T.Baker® BAKERBOND spe™, Filtration Columns with Frits
Supplier: Avantor
J.T.Baker® BAKERBOND spe™
7121-06EA 113 EUR
7121-06 7121-03 7121-01
J.T.Baker® BAKERBOND spe™, Filtration Columns with Frits
Chromatography Columns
These empty filtration SPE columns are packed with dual 20 µm frits. They are designed for use with spe-12G and spe-24G glass column processors.