Tetrasodium diphosphate decahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Sodium pyrophosphate decahydrate, tetra-Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, TSPP
M-Clarity™ quality level = MQ200
S6422-100G S6422-500G
SIALS6422-100GEA 55.4 EUR
SIALS6422-100G SIALS6422-500G
Tetrasodium diphosphate decahydrate, Sigma-Aldrich®
tetra-Sodium diphosphate decahydrate
Formula: Na₄P₂O₇·10H₂O
MW: 446.06 g/mol
Melting Pt: 990
Density: 1.82 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149200
CAS Number: 13472-36-1
EINECS: 231-767-1