Trisodium phosphate dodecahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
S7778-1KG S7778-500G
SIALS7778-1KGEA 186 EUR
SIALS7778-1KG SIALS7778-500G
Trisodium phosphate dodecahydrate, Sigma-Aldrich®
tri-Sodium phosphate dodecahydrate
Formula: Na₃PO₄·12H₂O
Melting Pt: ∼75 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149198
CAS Number: 10101-89-0
EINECS: 231-509-8
ADR: 8,III
REACH: 01-2119489800-32-xxxx
Merck Index: 13,08736