Sodium thiosulphate pentahydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
S6672-1KG S6672-500G
SIALS6672-1KGEA 187 EUR
SIALS6672-1KG SIALS6672-500G
Sodium thiosulphate pentahydrate, Sigma-Aldrich®
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Melting Pt: 48
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844