Calcium nitrate tetrahydrate
Supplier: Roth Carl
Warning
X886.1 X886.2 X886.3
ROTHX886.1EA 19.5 EUR
ROTHX886.1 ROTHX886.2 ROTHX886.3
Calcium nitrate tetrahydrate
Calcium nitrate tetrahydrate
Formula: Ca(NO₃)₂·4H₂O
MW: 236.15 g/mol
Melting Pt: ∼42 °C
Density: 1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4
EINECS: 233-332-1
UN: 1454
ADR: 5.1 III
Merck Index: 13,01687