Calcium nitrate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
C2786-1KG C2786-500G
SIALC2786-1KGEA 209 EUR
SIALC2786-1KG SIALC2786-500G
Calcium nitrate tetrahydrate, Sigma-Aldrich®
Calcium nitrate tetrahydrate
Formula: Ca(NO₃)₂·4H₂O
MW: 236.15 g/mol
Melting Pt: ∼42 °C
Density: 1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4
EINECS: 233-332-1
UN: 1454
ADR: 5.1 III
Merck Index: 13,01687